Assay Details
| Gene Symbol: METAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126711.6 | XP_003126759.3 | 690 | 93 | ||
| XM_005664247.3 | XP_005664304.1 | 686 | 93 | |||
| GenBank mRNA | AK236078.1 | - | 5 - 6 | 675 | 93 | |
| AK239778.1 | - | 5 - 6 | 688 | 93 | ||
| AK395008.1 | - | 5 - 6 | 761 | 93 | ||
More Information
Set Membership: |
|

METAP2
Genomic Map