Assay Details
| Gene Symbol: ABCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123069.1 | NP_001116541.1 | 15 - 16 | 1486 | 63 | |
| XM_005653453.3 | XP_005653510.1 | 1677 | 63 | |||
| XM_005653454.3 | XP_005653511.1 | 1674 | 63 | |||
More Information
Set Membership: |
|

ABCF1
Genomic Map