Assay Details
| Gene Symbol: UBE2Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929469.6 | XP_001929504.3 | 1364 | 66 | ||
| GenBank mRNA | AK237001.1 | - | 10 - 11 | 791 | 66 | |
| AY609725.1 | - | 12 - 13 | 1137 | 66 | ||
More Information
Set Membership: |
|

UBE2Q1
Genomic Map