Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244335.1 | NP_001231264.1 | 1 - 2 | 226 | 91 | |
GenBank mRNA | AK392453.1 | - | 1 - 2 | 174 | 92 | |
AK399915.1 | - | 1 - 2 | 134 | 91 | ||
AK400150.1 | - | 1 - 2 | 188 | 91 | ||
BE032382.1 | - | 1 - 2 | 227 | 91 | ||
FS666793.1 | - | 1 - 2 | 102 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244335.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
