Assay Details
Gene Symbol: TMEM184C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021101527.1 | XP_020957186.1 | 1510 | 76 | ||
GenBank mRNA | AK233425.1 | - | 8 - 9 | 1424 | 76 | |
AK391729.1 | - | 8 - 9 | 1436 | 76 | ||
AK395561.1 | - | 8 - 9 | 1070 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
