Assay Details
| Gene Symbol: SMARCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134577.6 | XP_003134625.3 | 893 | 83 | ||
| XM_013990738.2 | XP_013846192.1 | 316 | 83 | |||
| XM_021079264.1 | XP_020934923.1 | 620 | 83 | |||
| XM_021079265.1 | XP_020934924.1 | 897 | 83 | |||
| XM_021079266.1 | XP_020934925.1 | 619 | 83 | |||
| GenBank mRNA | AK349383.1 | - | 5 - 6 | 605 | 83 | |
More Information
Set Membership: |
|

SMARCD3
Genomic Map