Assay Details
| Gene Symbol: ARPC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134355.1 | NP_001127827.1 | 3 - 4 | 391 | 85 | |
| XM_005661855.3 | XP_005661912.1 | 573 | 85 | |||
| GenBank mRNA | AK232038.1 | - | 4 - 5 | 553 | 85 | |
| EU868833.1 | - | 3 - 4 | 391 | 85 | ||
More Information
Set Membership: |
|

ARPC1A
Genomic Map