Assay Details
| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074959.1 | XP_020930618.1 | 1507 | 84 | ||
| XM_021074960.1 | XP_020930619.1 | 1473 | 84 | |||
| GenBank mRNA | AK395088.1 | - | 1 - 2 | 121 | 84 | |
More Information
Set Membership: |
|

NCL
Genomic Map