Assay Details
| Gene Symbol: RAB6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243273.1 | NP_001230202.1 | 2 - 3 | 542 | 98 | |
| XM_021102115.1 | XP_020957774.1 | 104 | 98 | |||
| GenBank mRNA | AK232771.1 | - | 2 - 3 | 542 | 98 | |
| AK346415.1 | - | 3 - 4 | 458 | 98 | ||
| AY609534.1 | - | 2 - 3 | 277 | 98 | ||
More Information
Set Membership: |
|

RAB6A
Genomic Map