Assay Details
| Gene Symbol: RBM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359964.4 | XP_003360012.1 | 1922 | 148 | ||
| XM_003359965.4 | XP_003360013.1 | 1672 | 148 | |||
| XM_005672897.3 | XP_005672954.1 | 1763 | 148 | |||
| XM_005672898.3 | XP_005672955.1 | 1689 | 148 | |||
| XM_013985327.2 | XP_013840781.1 | 1678 | 148 | |||
| XM_021078267.1 | XP_020933926.1 | 1805 | 148 | |||
| XM_021078268.1 | XP_020933927.1 | 3275 | 148 | |||
| XM_021078269.1 | XP_020933928.1 | 2517 | 148 | |||
| XM_021078270.1 | XP_020933929.1 | 1806 | 148 | |||
| XM_021078271.1 | XP_020933930.1 | 3304 | 148 | |||
| XM_021078272.1 | XP_020933931.1 | 4363 | 148 | |||
| XM_021078273.1 | XP_020933932.1 | 1793 | 148 | |||
| XM_021078274.1 | XP_020933933.1 | 1872 | 148 | |||
| XM_021078275.1 | XP_020933934.1 | 2902 | 148 | |||
| XM_021078276.1 | XP_020933935.1 | 2172 | 148 | |||
| XM_021078278.1 | XP_020933937.1 | 3179 | 148 | |||
| XM_021078279.1 | XP_020933938.1 | 2451 | 148 | |||
| XM_021078280.1 | XP_020933939.1 | 1767 | 148 | |||
| XM_021078281.1 | XP_020933940.1 | 3444 | 148 | |||
| XM_021078282.1 | XP_020933941.1 | 4269 | 148 | |||
| XM_021078283.1 | XP_020933942.1 | 4995 | 148 | |||
| GenBank mRNA | AM492787.1 | - | 4 - 5 | 312 | 148 | |
More Information
Set Membership: |
|

RBM39
Genomic Map