Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043346.1 | NP_001036811.1 | 7 - 8 | 1310 | 77 | |
| GenBank mRNA | DQ307487.1 | - | 6 - 7 | 928 | 77 | |
| DQ321701.1 | - | 7 - 8 | 1310 | 77 | ||
More Information
Set Membership: |
|

HCRTR1
Genomic Map