Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214149.1 | NP_999314.1 | 3 - 4 | 579 | 79 | |
| GenBank mRNA | AF220248.1 | - | 3 - 4 | 579 | 79 | |
| AF281633.1 | - | 3 - 4 | 566 | 79 | ||
More Information
Set Membership: |
|

CTLA4
Genomic Map