Assay Details
| Gene Symbol: C4H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204390.1 | NP_001191319.1 | 6 - 7 | 827 | 78 | |
| GenBank mRNA | AK231836.1 | - | 5 - 6 | 586 | 78 | |
| AK233670.1 | - | 6 - 7 | 795 | 78 | ||
| AK234116.1 | - | 6 - 7 | 795 | 78 | ||
| AK351176.1 | - | 6 - 7 | 722 | 78 | ||
| AK397935.1 | - | 6 - 7 | 870 | 78 | ||
| AY609895.1 | - | 6 - 7 | 720 | 78 | ||
| EU814887.1 | - | 6 - 7 | 796 | 78 | ||
More Information
Set Membership: |
|

C4H1orf43
Genomic Map