Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244917.1 | NP_001231846.1 | 3 - 4 | 198 | 97 | |
| XM_005665239.3 | XP_005665296.2 | 308 | 97 | |||
| GenBank mRNA | AK231118.1 | - | 3 - 4 | 202 | 97 | |
| AK236342.1 | - | 3 - 4 | 198 | 97 | ||
| AK344747.1 | - | 3 - 4 | 173 | 97 | ||
| AY610032.1 | - | 3 - 4 | 174 | 97 | ||
More Information
Set Membership: |
|

MYCBP
Genomic Map