Assay Details
| Gene Symbol: DLX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159660.1 | NP_001153132.1 | 1 - 2 | 352 | 64 | |
| GenBank mRNA | AK240383.1 | - | 1 - 2 | 547 | 64 | |
More Information
Set Membership: |
|

DLX5
Genomic Map