Assay Details
| Gene Symbol: PPP1CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044560.1 | NP_001038025.1 | 4 - 5 | 521 | 80 | |
| GenBank mRNA | AK235498.1 | - | 4 - 5 | 753 | 80 | |
| AK392314.1 | - | 4 - 5 | 743 | 80 | ||
| DQ380141.1 | - | 4 - 5 | 521 | 80 | ||
More Information
Set Membership: |
|

PPP1CC
Genomic Map