Assay Details
| Gene Symbol: NME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244449.1 | NP_001231378.1 | 4 - 5 | 423 | 53 | |
| NM_001244450.1 | NP_001231379.1 | 4 - 5 | 420 | 53 | ||
| NR_045137.1 | 3 - 4 | 475 | 53 | |||
| GenBank mRNA | AK240056.1 | - | 3 - 4 | 471 | 53 | |
| AK394553.1 | - | 3 - 4 | 474 | 53 | ||
| BF080492.1 | - | 4 - 5 | 423 | 53 | ||
| BW980049.1 | - | 4 - 5 | 392 | 53 | ||
| CJ014819.1 | - | 3 - 4 | 475 | 53 | ||
| EW092573.2 | - | 1 - 2 | 89 | 53 | ||
More Information
Set Membership: |
|

NME3
Genomic Map