Assay Details
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095332.1 | XP_020950991.1 | 916 | 66 | ||
GenBank mRNA | AK392557.1 | - | 4 - 5 | 646 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
