Assay Details
| Gene Symbol: PTGDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122669.4 | XP_003122717.1 | 1251 | 63 | ||
| XM_021082964.1 | XP_020938623.1 | 1345 | 63 | |||
| GenBank mRNA | AK392083.1 | - | 2 | 538 | 63 | |
More Information
Set Membership: |
|

PTGDR2
Genomic Map