Assay Details
| Gene Symbol: CASQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089995.1 | XP_020945654.1 | 901 | 85 | ||
| GenBank mRNA | AF034612.1 | - | 3 - 4 | 284 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CASQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089995.1 | XP_020945654.1 | 901 | 85 | ||
| GenBank mRNA | AF034612.1 | - | 3 - 4 | 284 | 85 | |
Set Membership: |
|