Assay Details
| Gene Symbol: MMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166308.1 | NP_001159780.1 | 7 | 1081 | 119 | |
| GenBank mRNA | AF069641.1 | - | 1 | 50 | 119 | |
| AK231271.1 | - | 7 | 1087 | 119 | ||
| AK391220.1 | - | 7 | 1087 | 119 | ||
| EU722907.1 | - | 7 | 1081 | 119 | ||
More Information
Set Membership: |
|

MMP3
Genomic Map