Assay Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002817.1 | NP_001002817.1 | 2 - 3 | 314 | 72 | |
| GenBank mRNA | AF102872.1 | - | 1 - 2 | 166 | 72 | |
| AJ416020.1 | - | 2 - 3 | 244 | 72 | ||
| AK344105.1 | - | 2 - 3 | 312 | 72 | ||
| AY610297.1 | - | 2 - 3 | 298 | 72 | ||
| HM453202.1 | - | 2 - 3 | 254 | 72 | ||
More Information
Set Membership: |
|

FABP4
Genomic Map