Assay Details
| Gene Symbol: SLC9A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354711.3 | XP_003354759.1 | 2111 | 89 | ||
| GenBank mRNA | AF124218.1 | - | 1 - 2 | 27 | 89 | |
More Information
Set Membership: |
|

SLC9A4
Genomic Map