Assay Details
| Gene Symbol: TPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129947.1 | NP_001123419.1 | 7 - 8 | 814 | 63 | |
| XM_005660207.3 | XP_005660264.1 | 893 | 63 | |||
| XM_005660208.3 | XP_005660265.1 | 893 | 63 | |||
| XM_021080975.1 | XP_020936634.1 | 1579 | 63 | |||
| GenBank mRNA | AF420022.1 | - | 5 - 6 | 427 | 63 | |
| AK234959.1 | - | 7 - 8 | 749 | 63 | ||
| AK237158.1 | - | 7 - 8 | 890 | 63 | ||
| AK345719.1 | - | 7 - 8 | 806 | 63 | ||
| AK394427.1 | - | 7 - 8 | 817 | 63 | ||
| AK394492.1 | - | 7 - 8 | 817 | 63 | ||
| AK394663.1 | - | 7 - 8 | 815 | 63 | ||
| AK394755.1 | - | 7 - 8 | 743 | 63 | ||
| AK394995.1 | - | 7 - 8 | 894 | 63 | ||
| AK397959.1 | - | 7 - 8 | 893 | 63 | ||
| AK398530.1 | - | 7 - 8 | 818 | 63 | ||
| AK399619.1 | - | 7 - 8 | 696 | 63 | ||
| DQ306707.1 | - | 7 - 8 | 814 | 63 | ||
More Information
Set Membership: |
|

TPM2
Genomic Map