Assay Details
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243346.1 | NP_001230275.2 | 2 | 988 | 92 | |
| NM_001272094.1 | NP_001259023.1 | 2 | 1000 | 92 | ||
| XM_013997805.2 | XP_013853259.1 | 1389 | 92 | |||
| XM_021090916.1 | XP_020946575.1 | 1372 | 92 | |||
| GenBank mRNA | AJ291440.1 | - | 1 | 547 | 92 | |
| AK348447.1 | - | 2 | 982 | 92 | ||
| AK350179.1 | - | 2 | 897 | 92 | ||
| AK395621.1 | - | 2 | 988 | 92 | ||
More Information
Set Membership: |
|

ART4
Genomic Map