Assay Details
| Gene Symbol: LOC396685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978400.2 | XP_013833854.1 | 819 | 89 | ||
| GenBank mRNA | AJ293891.1 | - | 1 - 2 | 72 | 89 | |
| AK232550.1 | - | 2 - 3 | 742 | 89 | ||
| AK398935.1 | - | 2 - 3 | 719 | 89 | ||
More Information
Set Membership: |
|

LOC396685
Genomic Map