Assay Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081549.1 | XP_020937208.1 | 240 | 61 | ||
| XM_021081550.1 | XP_020937209.1 | 243 | 61 | |||
| XM_021081551.1 | XP_020937210.1 | 233 | 61 | |||
| XM_021081552.1 | XP_020937211.1 | 426 | 61 | |||
| XM_021081553.1 | XP_020937212.1 | 269 | 61 | |||
| XM_021081554.1 | XP_020937213.1 | 276 | 61 | |||
| XM_021081555.1 | XP_020937214.1 | 240 | 61 | |||
| XM_021081556.1 | XP_020937215.1 | 241 | 61 | |||
| GenBank mRNA | AJ539380.1 | - | 128 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021081552.1, XM_021081553.1, XM_021081554.1 transcripts.
Set Membership: |
|

NUDT14
Genomic Map