Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128456.1 | NP_001121928.1 | 1 - 2 | 96 | 95 | |
| XM_005655480.3 | XP_005655537.1 | 426 | 95 | |||
| XM_005655481.3 | XP_005655538.1 | 536 | 95 | |||
| GenBank mRNA | AK240087.1 | - | 2 - 3 | 413 | 95 | |
| AK398481.1 | - | 2 - 3 | 409 | 95 | ||
| EU366153.1 | - | 1 - 2 | 96 | 95 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map