Assay Details
| Gene Symbol: SLC1A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996100.2 | XP_013851554.1 | 2272 | 89 | ||
| XM_021087450.1 | XP_020943109.1 | 2029 | 89 | |||
| GenBank mRNA | AK394839.1 | - | 6 - 7 | 681 | 89 | |
| AY375264.1 | - | 1 - 2 | 36 | 89 | ||
More Information
Set Membership: |
|

SLC1A4
Genomic Map