Assay Details
| Gene Symbol: RXRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130213.1 | NP_001123685.1 | 6 - 7 | 1079 | 60 | |
| XM_005655370.3 | XP_005655427.1 | 855 | 60 | |||
| XM_013988076.2 | XP_013843530.1 | 1737 | 60 | |||
| XM_021088737.1 | XP_020944396.1 | 1132 | 60 | |||
| GenBank mRNA | AB530150.1 | - | 4 - 5 | 540 | 60 | |
| AK237831.1 | - | 5 - 6 | 835 | 60 | ||
| AK394594.1 | - | 5 - 6 | 840 | 60 | ||
| AY429474.1 | - | 5 - 6 | 814 | 60 | ||
| DQ866834.1 | - | 6 - 7 | 1079 | 60 | ||
More Information
Set Membership: |
|

RXRG
Genomic Map