Assay Details
| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161753.1 | NP_001155225.1 | 4 - 5 | 843 | 68 | |
| XM_021098318.1 | XP_020953977.1 | 733 | 68 | |||
| GenBank mRNA | AK233595.1 | - | 4 - 5 | 765 | 68 | |
| AK394136.1 | - | 4 - 5 | 765 | 68 | ||
| AY509878.1 | - | 1 - 2 | 193 | 68 | ||
More Information
Set Membership: |
|

PCK2
Genomic Map