Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091712.1 | XP_020947371.1 | 1725 | 80 | ||
| XM_021091714.1 | XP_020947373.1 | 2490 | 80 | |||
| XM_021091715.1 | XP_020947374.1 | 1671 | 80 | |||
| GenBank mRNA | AY514037.1 | - | 1 | 47 | 80 | |
More Information
Set Membership: |
|

SP7
Genomic Map