Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064314.1 | XP_020919973.1 | 4479 | 94 | ||
| XM_021064315.1 | XP_020919974.1 | 4481 | 94 | |||
| GenBank mRNA | AK231190.1 | - | 5 - 6 | 743 | 94 | |
| AK238970.1 | - | 12 - 13 | 1760 | 94 | ||
| AK346654.1 | - | 4 - 5 | 590 | 94 | ||
| AK389203.1 | - | 13 - 14 | 1882 | 94 | ||
| AY706381.2 | - | 5 - 6 | 771 | 94 | ||
More Information
Set Membership: |
|

EPRS1
Genomic Map