Assay Details
| Gene Symbol: IL23R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137621.1 | NP_001131093.1 | 3 | 261 | 127 | |
| XM_021093601.1 | XP_020949260.1 | 1370 | 127 | |||
| GenBank mRNA | AK344335.1 | - | 2 | 1419 | 127 | |
| AY876385.1 | - | 1 | 68 | 127 | ||
| AY948114.1 | - | 3 | 261 | 127 | ||
More Information
Set Membership: |
|

IL23R
Genomic Map