Assay Details
| Gene Symbol: IL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091967.1 | XP_020947626.1 | 487 | 70 | ||
| XM_021091968.1 | XP_020947627.1 | 487 | 70 | |||
| GenBank mRNA | AY937228.1 | - | 1 - 2 | 150 | 70 | |
| KX588234.1 | - | 3 - 4 | 425 | 70 | ||
More Information
Set Membership: |
|

IL22
Genomic Map