Assay Details
| Gene Symbol: SLC16A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128445.1 | NP_001121917.1 | 3 - 4 | 1257 | 115 | |
| XM_021088450.1 | XP_020944109.1 | 1558 | 115 | |||
| XM_021088451.1 | XP_020944110.1 | 1923 | 115 | |||
| GenBank mRNA | AK238822.1 | - | 4 - 5 | 1432 | 115 | |
| DQ885287.1 | - | 1 - 2 | 563 | 115 | ||
| EU404088.1 | - | 1 - 2 | 655 | 115 | ||
| EU559708.1 | - | 3 - 4 | 1257 | 115 | ||
More Information
Set Membership: |
|

SLC16A1
Genomic Map