Assay Details
| Gene Symbol: KCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092437.1 | XP_020948096.1 | 2861 | 72 | ||
| GenBank mRNA | EF424223.1 | - | 1 | 551 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092437.1 | XP_020948096.1 | 2861 | 72 | ||
| GenBank mRNA | EF424223.1 | - | 1 | 551 | 72 | |
Set Membership: |
|