Assay Details
| Gene Symbol: TNFRSF11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481346.4 | XP_003481394.3 | 613 | 75 | ||
| GenBank mRNA | AK390399.1 | - | 2 - 3 | 349 | 75 | |
| EF543195.2 | - | 1 - 2 | 333 | 75 | ||
More Information
Set Membership: |
|

TNFRSF11B
Genomic Map