Assay Details
| Gene Symbol: F11R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128444.1 | NP_001121916.1 | 5 - 6 | 621 | 79 | |
| GenBank mRNA | AK390649.1 | - | 5 - 6 | 678 | 79 | |
| EU056579.1 | - | 1 - 2 | 166 | 79 | ||
| EU670046.1 | - | 5 - 6 | 621 | 79 | ||
More Information
Set Membership: |
|

F11R
Genomic Map