Assay Details
| Gene Symbol: LOC100736733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315796.1 | NP_001302725.1 | 1 - 2 | 78 | 95 | |
| GenBank mRNA | AK231223.1 | - | 2 - 3 | 222 | 95 | |
| AK392957.1 | - | 2 - 3 | 224 | 95 | ||
| AY609994.1 | - | 2 - 3 | 208 | 95 | ||
| EU560428.1 | - | 1 - 2 | 78 | 95 | ||
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128453.1 | NP_001121925.1 | 3 - 4 | 222 | 95 | |
| GenBank mRNA | AK231394.1 | - | 2 - 3 | 221 | 95 | |
| Gene Symbol: LOC110255228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089549.1 | XP_020945208.1 | 227 | 95 | ||
| Gene Symbol: LOC110260309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089551.1 | XP_020945210.1 | 233 | 95 | ||
| Gene Symbol: LOC110260310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089553.1 | XP_020945212.1 | 232 | 95 | ||
| GenBank mRNA | EU652941.1 | - | 3 - 4 | 222 | 95 | |
More Information
Set Membership: |
|

LOC100736733
Genomic Map