Assay Details
| Gene Symbol: LOC100736733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315796.1 | NP_001302725.1 | 2 - 3 | 194 | 74 | |
| XM_021088677.1 | XP_020944336.1 | 351 | 74 | |||
| GenBank mRNA | AY609994.1 | - | 3 - 4 | 324 | 74 | |
| EU560428.1 | - | 2 - 3 | 194 | 74 | ||
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128453.1 | NP_001121925.1 | 4 - 5 | 338 | 74 | |
| Gene Symbol: LOC110260310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU652941.1 | - | 4 - 5 | 338 | 74 | |
More Information
Set Membership: |
|

LOC100736733
Genomic Map