Assay Details
| Gene Symbol: LOC100736733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315796.1 | NP_001302725.1 | 3 - 4 | 434 | 88 | |
| XM_021088677.1 | XP_020944336.1 | 591 | 88 | |||
| GenBank mRNA | AY609994.1 | - | 4 - 5 | 564 | 88 | |
| EU560428.1 | - | 3 - 4 | 434 | 88 | ||
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128453.1 | NP_001121925.1 | 5 - 6 | 578 | 88 | |
| XM_013996842.2 | XP_013852296.1 | 590 | 88 | |||
| GenBank mRNA | AK231394.1 | - | 4 - 5 | 577 | 88 | |
| Gene Symbol: LOC110255228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089549.1 | XP_020945208.1 | 583 | 88 | ||
| XM_021089550.1 | XP_020945209.1 | 580 | 88 | |||
| Gene Symbol: LOC110260310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089553.1 | XP_020945212.1 | 588 | 88 | ||
| XM_021089554.1 | XP_020945213.1 | 586 | 88 | |||
| GenBank mRNA | EU652941.1 | - | 5 - 6 | 578 | 88 | |
More Information
Set Membership: |
|

LOC100736733
Genomic Map