Assay Details
| Gene Symbol: DLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128466.1 | NP_001121938.1 | 3 - 4 | 267 | 109 | |
| XM_005666030.3 | XP_005666087.1 | 457 | 109 | |||
| XM_021097971.1 | XP_020953630.1 | 641 | 109 | |||
| GenBank mRNA | AK350063.1 | - | 4 - 5 | 428 | 109 | |
| EU589332.1 | - | 3 - 4 | 267 | 109 | ||
More Information
Set Membership: |
|

DLK2
Genomic Map