Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660371.3 | XP_005660428.1 | 378 | 61 | ||
| XM_005660372.3 | XP_005660429.1 | 480 | 61 | |||
| XM_021067967.1 | XP_020923626.1 | 377 | 61 | |||
| GenBank mRNA | AK233369.1 | - | 3 - 4 | 363 | 61 | |
| AK346200.1 | - | 3 - 4 | 363 | 61 | ||
| AK346379.1 | - | 3 - 4 | 363 | 61 | ||
| M35990.1 | - | 3 - 4 | 269 | 61 | ||
More Information
Set Membership: |
|

ORM1
Genomic Map