Assay Details
| Gene Symbol: LDLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206354.2 | NP_001193283.1 | 17 | 2601 | 78 | |
| XM_021080444.1 | XP_020936103.1 | 2808 | 78 | |||
| XM_021080449.1 | XP_020936108.1 | 2706 | 78 | |||
| XM_021080452.1 | XP_020936111.1 | 2735 | 78 | |||
| XM_021080457.1 | XP_020936116.1 | 3416 | 78 | |||
| GenBank mRNA | AF065990.1 | - | 16 | 2363 | 78 | |
| AF067952.1 | - | 16 | 2363 | 78 | ||
| AF118147.1 | - | 16 | 2306 | 78 | ||
| AK236400.1 | - | 8 | 986 | 78 | ||
| KF717533.1 | - | 17 | 2601 | 78 | ||
| S64272.1 | - | 1 | 71 | 78 | ||
More Information
Set Membership: |
|

LDLR
Genomic Map