Assay Details
| Gene Symbol: TBXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074772.1 | XP_020930431.1 | 668 | 74 | ||
| XM_021074778.1 | XP_020930437.1 | 668 | 74 | |||
| GenBank mRNA | U91519.1 | - | 1 - 2 | 43 | 74 | |
More Information
Set Membership: |
|

TBXT
Genomic Map