Assay Details
| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096908.1 | XP_020952567.1 | 1632 | 79 | ||
| GenBank mRNA | KU672521.1 | - | 8 | 1148 | 79 | |
| X17058.1 | - | 1253 | 79 | |||
More Information
Set Membership: |
|

SLC2A1
Genomic Map