Assay Details
| Gene Symbol: CNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353959.3 | XP_003354007.2 | 537 | 71 | ||
| GenBank mRNA | AK234333.1 | - | 5 | 515 | 71 | |
| AK397963.1 | - | 2 | 157 | 71 | ||
| Z19539.1 | - | 453 | 71 | |||
More Information
Set Membership: |
|

CNN2
Genomic Map