Assay Details
| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161637.1 | NP_001155109.1 | 1 | 509 | 73 | |
| XM_005661969.2 | XP_005662026.1 | 636 | 73 | |||
| XM_013995522.2 | XP_013850976.1 | 627 | 73 | |||
| XM_021085910.1 | XP_020941569.1 | 610 | 73 | |||
| GenBank mRNA | AB235916.1 | - | 1 | 145 | 73 | |
| AK233156.1 | - | 1 | 567 | 73 | ||
| AK393993.1 | - | 1 | 567 | 73 | ||
| FJ873111.1 | - | 1 | 509 | 73 | ||
More Information
Set Membership: |
|

CLDN4
Genomic Map