Assay Details
| Gene Symbol: LOC100623720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071094.1 | XP_020926753.1 | 840 | 61 | ||
| XM_021071095.1 | XP_020926754.1 | 653 | 61 | |||
| XM_021071096.1 | XP_020926755.1 | 653 | 61 | |||
| GenBank mRNA | AB237776.1 | - | 1 | 53 | 61 | |
More Information
Set Membership: |
|

LOC100623720
Genomic Map